Assay Details
Target Gene Details
Entrez Gene ID: | 83698 |
Gene Name: | calneuron 1 |
Gene Aliases: |
CABP8 |
Location: |
Chr.7:71779491-72504271 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALN1 | NM_001017440.2 | 6 | 5030 | NP_001017440.1 |
| NM_031468.3 | 7 | 5020 | NP_113656.2 | |
| XM_011516594.2 | 6 | 4872 | XP_011514896.1 | |
| XM_011516595.1 | 6 | 4620 | XP_011514897.1 | |
| XM_011516596.2 | 7 | 5515 | XP_011514898.1 | |
| XM_011516597.2 | 6 | 5448 | XP_011514899.1 | |
| XM_011516599.1 | 6 | 4819 | XP_011514901.1 | |
| XM_017012675.1 | 8 | 5375 | XP_016868164.1 | |
| XM_017012676.1 | 7 | 5157 | XP_016868165.1 | |
| XM_017012677.1 | 6 | 4670 | XP_016868166.1 | |
| XM_017012678.1 | 7 | 5132 | XP_016868167.1 | |
| XM_017012679.1 | 6 | 5485 | XP_016868168.1 | |
| XM_017012680.1 | 6 | 4784 | XP_016868169.1 | |
| XM_017012681.1 | 6 | 4647 | XP_016868170.1 | |
| XM_017012682.1 | 6 | 5197 | XP_016868171.1 | |
| XM_017012683.1 | 7 | 4981 | XP_016868172.1 | |
| AF282250.1 | 7 | 4751 | AAK15155.1 | |
| AK094469.1 | 1 | 988 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516049 | Chr.7:71561478 - 71815314 on Build GRCh38 | Gain |
|
| nsv607459 | Chr.7:71774529 - 71801548 on Build GRCh38 | Gain |
|
| esv3613640 | Chr.7:71778998 - 71802508 on Build GRCh38 | Gain |
|
| nsv1033922 | Chr.7:71750208 - 71802242 on Build GRCh38 | Gain |
|
| nsv1017575 | Chr.7:70687626 - 71905471 on Build GRCh38 | Gain |
|
| dgv6456n100 | Chr.7:71568840 - 71823976 on Build GRCh38 | Gain |
|
| nsv1026896 | Chr.7:71538477 - 72256531 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map