Assay Details
Target Gene Details
Entrez Gene ID: | 8802 |
Gene Name: | succinate-CoA ligase alpha subunit |
Gene Aliases: |
GALPHA, MTDPS9, SUCLA1 |
Location: |
Chr.2:84423523-84459462 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUCLG1 | NM_003849.3 | 2 | 338 | NP_003840.2 |
| AF104921.2 | 2 | 137 | AAD17940.2 | |
| AK225737.1 | 2 | 115 | ||
| AK291082.1 | 2 | 137 | ||
| AK296741.1 | 2 | 145 | ||
| AK313290.1 | 2 | 117 | ||
| BC000504.2 | 2 | 141 | AAH00504.1 | |
| BM804835.1 | 2 | 324 | ||
| BP194715.1 | 2 | 339 | ||
| CR457139.1 | 2 | 106 | CAG33420.1 | |
| Z68204.1 | 2 | 112 | CAA92426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002072 | Chr.2:84218779 - 84482091 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map