Assay Details
Target Gene Details
Entrez Gene ID: | 338692 |
Gene Name: | ankyrin repeat domain 13D |
Gene Aliases: |
- |
Location: |
Chr.11:67289291-67302484 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13D | NM_207354.2 | 2 | 357 | NP_997237.2 |
| NR_030767.1 | 3 | 440 | ||
| XM_011544985.1 | 2 | 300 | XP_011543287.1 | |
| XM_011544986.1 | 3 | 440 | XP_011543288.1 | |
| XM_011544987.2 | 2 | 260 | XP_011543289.1 | |
| AK096277.1 | 2 | 270 | ||
| AK124721.1 | 3 | 437 | BAC85932.1 | |
| AK126438.1 | 2 | 246 | BAC86550.1 | |
| BC110419.1 | 2 | 330 | ||
| BC121024.1 | 2 | 231 | ||
| BC121025.2 | 2 | 231 | ||
| DA870287.1 | 3 | 440 | ||
| DB460018.1 | 2 | 357 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555253 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
| nsv951021 | Chr.11:67283730 - 67292729 on Build GRCh38 | Deletion |
|

Genomic Map