Assay Details
Target Gene Details
Entrez Gene ID: | 55421 |
Gene Name: | nuclear cap binding subunit 3 |
Gene Aliases: |
C17orf85, ELG, HSA277841 |
Location: |
Chr.17:3806748-3846251 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCBP3 | NM_001114118.2 | 13 | 3239 | NP_001107590.1 |
| XM_005256717.2 | 14 | 2777 | XP_005256774.1 | |
| XM_006721556.2 | 13 | 2666 | XP_006721619.1 | |
| XM_011523954.2 | 14 | 2694 | XP_011522256.1 | |
| XM_011523955.2 | 13 | 2548 | XP_011522257.1 | |
| XM_017024826.1 | XP_016880315.1 | |||
| XM_017024827.1 | 8 | 2251 | XP_016880316.1 | |
| XM_017024828.1 | 9 | 2069 | XP_016880317.1 | |
| XM_017024829.1 | XP_016880318.1 | |||
| AJ277841.1 | 8 | 2536 | CAB91001.1 | |
| AK125048.1 | 10 | 3928 | ||
| DA855232.1 | 1 | 71 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| esv2761940 | Chr.17:3767589 - 3997617 on Build GRCh38 | Gain |
|
| nsv469791 | Chr.17:3700760 - 3885899 on Build GRCh38 | Loss |
|
| nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map