Assay Details
Target Gene Details
Entrez Gene ID: | 123879 |
Gene Name: | defective in cullin neddylation 1 domain containing 3 |
Gene Aliases: |
44M2.4, SCCRO3 |
Location: |
Chr.16:20854925-20900358 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCUN1D3 | NM_173475.3 | NP_775746.1 | ||
| BC040442.1 | AAH40442.1 | |||
| BX648998.1 | ||||
| DA545688.1 |
Target Gene Details
Entrez Gene ID: | 57149 |
Gene Name: | LYR motif containing 1 |
Gene Aliases: |
A211C6.1 |
Location: |
Chr.16:20899626-20925006 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYRM1 | NM_020424.4 | NP_065157.1 | ||
| XM_011545902.1 | XP_011544204.1 | |||
| XM_011545908.2 | XP_011544210.1 | |||
| XM_017023475.1 | 2 | 440 | XP_016878964.1 | |
| XM_017023476.1 | 2 | 446 | XP_016878965.1 | |
| XM_017023477.1 | 2 | 449 | XP_016878966.1 | |
| XM_017023480.1 | XP_016878969.1 | |||
| AK308690.1 | ||||
| DA572182.1 | ||||
| DB474686.1 | 2 | 208 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map