Assay Details
Target Gene Details
Entrez Gene ID: | 1573 |
Gene Name: | cytochrome P450 family 2 subfamily J member 2 |
Gene Aliases: |
CPJ2, CYPIIJ2 |
Location: |
Chr.1:59893308-59926798 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2J2 | NM_000775.3 | 9 | 1471 | NP_000766.2 |
| NR_134981.1 | 8 | 1283 | ||
| NR_134982.1 | 10 | 1622 | ||
| AB080265.1 | 9 | 1424 | BAB85489.1 | |
| AK055088.1 | 8 | 2125 | ||
| AK315387.1 | 9 | 1439 | ||
| BC032594.1 | 9 | 1474 | AAH32594.1 | |
| CD013972.1 | 8 | 1246 | ||
| CD013973.1 | 10 | 1585 | ||
| U37143.2 | 9 | 1424 | AAC50370.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012703 | Chr.1:59796670 - 59897484 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map