Assay Details
Target Gene Details
Entrez Gene ID: | 51503 |
Gene Name: | CWC15 spliceosome-associated protein |
Gene Aliases: |
AD002, C11orf5, Cwf15, HSPC148, ORF5 |
Location: |
Chr.11:94962622-94973612 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWC15 | NM_016403.3 | NP_057487.2 | ||
| XM_011542868.2 | XP_011541170.1 | |||
| XM_017017916.1 | XP_016873405.1 | |||
| AF110775.1 | AAF14858.1 | |||
| AF161497.1 | AAF29112.1 | |||
| AJ250393.1 | CAB96541.1 | |||
| AK225956.1 | ||||
| AK314900.1 | ||||
| BC040946.1 | AAH40946.1 | |||
| BM971024.1 | ||||
| BU676427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv446 | Chr.11:94960555 - 95005748 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map