Assay Details
Target Gene Details
Entrez Gene ID: | 9242 |
Gene Name: | musculin |
Gene Aliases: |
ABF-1, ABF1, MYOR, bHLHa22 |
Location: |
Chr.8:71841542-71844496 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSC | NM_005098.3 | 1 | 264 | NP_005089.2 |
| AF060154.1 | 1 | 158 | AAC15071.1 | |
| AK313102.1 | 1 | 221 | ||
| BC006313.2 | 1 | 230 | AAH06313.1 | |
| BC067827.1 | 1 | 239 | AAH67827.1 | |
| DB257094.1 | 1 | 264 |
Target Gene Details
Entrez Gene ID: | 100132891 |
Gene Name: | MSC antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:71843123-72056312 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSC-AS1 | NR_033651.1 | 1 | 118 | |
| NR_033652.1 | ||||
| AK123315.1 | ||||
| AK304427.1 | ||||
| BF171746.1 | 1 | 143 | ||
| BX647470.1 | 1 | 131 | ||
| DB161440.1 | 1 | 88 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831353 | Chr.8:71836557 - 72032198 on Build GRCh38 | Loss |
|
| esv33409 | Chr.8:71842768 - 71848488 on Build GRCh38 | Loss |
|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map