Assay Details
Target Gene Details
Entrez Gene ID: | 81553 |
Gene Name: | family with sequence similarity 49 member A |
Gene Aliases: |
- |
Location: |
Chr.2:16549459-16665866 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM49A | NM_030797.3 | 6 | 650 | NP_110424.1 |
| XM_005262630.3 | 5 | 606 | XP_005262687.1 | |
| XM_006712108.3 | 6 | 605 | XP_006712171.1 | |
| AK055334.1 | 6 | 618 | ||
| AK091280.1 | 1 | 64 | ||
| AK315065.1 | 5 | 545 | ||
| AL136704.1 | 6 | 615 | CAB66639.1 | |
| BC038971.1 | 6 | 601 | AAH38971.1 | |
| CR533504.1 | 4 | 397 | CAG38535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34115 | Chr.2:16338544 - 16692772 on Build GRCh38 | Loss |
|

Genomic Map