Assay Details
Target Gene Details
Entrez Gene ID: | 57107 |
Gene Name: | prenyl (decaprenyl) diphosphate synthase, subunit 2 |
Gene Aliases: |
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1 |
Location: |
Chr.6:107152557-107459683 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDSS2 | NM_020381.3 | 8 | 3278 | NP_065114.3 |
| XM_011535956.2 | 9 | 3574 | XP_011534258.1 | |
| XM_011535957.2 | 8 | 3452 | XP_011534259.1 | |
| XM_011535958.2 | 8 | 3440 | XP_011534260.1 | |
| XM_011535959.2 | 7 | 3219 | XP_011534261.1 | |
| XM_011535960.2 | 9 | 3203 | XP_011534262.1 | |
| XM_011535961.2 | 6 | 2992 | XP_011534263.1 | |
| XM_011535962.2 | 8 | 2920 | XP_011534264.1 | |
| AF254956.1 | 8 | 3249 | AAF97788.1 | |
| AJ420575.1 | 1 | 844 | ||
| AL832290.1 | 3 | 2083 | ||
| BC039906.1 | 8 | 3264 | AAH39906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6135n100 | Chr.6:106988892 - 107234753 on Build GRCh38 | Gain |
|
| dgv87n68 | Chr.6:106996980 - 107196871 on Build GRCh38 | Loss |
|
| esv2732546 | Chr.6:106816634 - 107364206 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map