Assay Details
Target Gene Details
Entrez Gene ID: | 80011 |
Gene Name: | family with sequence similarity 192 member A |
Gene Aliases: |
C16orf94, CDA018, CDA10, NIP30 |
Location: |
Chr.16:57152466-57186242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
More Information
Additional Information:
For this assay, SNP(s) [rs74544530] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map