Assay Details
Target Gene Details
Entrez Gene ID: | 25841 |
Gene Name: | ankyrin repeat and BTB domain containing 2 |
Gene Aliases: |
ABTB2A, BTBD22 |
Location: |
Chr.11:34150987-34358008 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABTB2 | NM_145804.2 | 17 | 4748 | NP_665803.2 |
| AK056863.1 | 5 | 1804 | ||
| AK091560.1 | 14 | 2966 | BAC03697.1 | |
| AK095632.1 | 17 | 3995 | BAC04595.1 | |
| AL050374.1 | 2 | 1520 | ||
| BX640841.1 | 9 | 2449 | CAE45913.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509400 | Chr.11:34110735 - 34232487 on Build GRCh38 | Insertion |
|
| nsv820477 | Chr.11:34150369 - 34152735 on Build GRCh38 | Duplication |
|
| esv3302725 | Chr.11:34150634 - 34152568 on Build GRCh38 | Tandem Duplication |
|
| dgv207n67 | Chr.11:34150481 - 34152529 on Build GRCh38 | Loss |
|
| esv26380 | Chr.11:34150545 - 34152640 on Build GRCh38 | Loss |
|
| nsv820233 | Chr.11:34149816 - 34152473 on Build GRCh38 | Loss |
|
| nsv951600 | Chr.11:34136054 - 34174953 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map