Assay Details
Target Gene Details
Entrez Gene ID: | 51699 |
Gene Name: | VPS29, retromer complex component |
Gene Aliases: |
DC15, DC7, PEP11 |
Location: |
Chr.12:110491523-110502140 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS29 | NM_001282150.1 | 6 | 988 | NP_001269079.1 |
| NM_001282151.1 | 3 | 597 | NP_001269080.1 | |
| NM_016226.4 | 4 | 833 | NP_057310.1 | |
| NM_057180.2 | 5 | 845 | NP_476528.1 | |
| NR_104099.1 | 6 | 885 | ||
| NR_104100.1 | 5 | 873 | ||
| NR_104101.1 | 5 | 923 | ||
| NR_104102.1 | 4 | 724 | ||
| XM_006719459.3 | 6 | 894 | XP_006719522.1 | |
| XM_006719460.3 | 7 | 931 | XP_006719523.1 | |
| XM_011538484.2 | 5 | 879 | XP_011536786.1 | |
| XM_017019457.1 | 5 | 904 | XP_016874946.1 | |
| XM_017019458.1 | 4 | 1024 | XP_016874947.1 | |
| XM_017019459.1 | 4 | 686 | XP_016874948.1 | |
| AF168716.1 | 5 | 803 | AAF87318.1 | |
| AF193795.1 | 4 | 795 | AAF04596.1 | |
| AK000499.1 | 3 | 561 | ||
| AK129767.1 | 4 | 795 | ||
| AL136614.1 | 4 | 811 | CAB66549.1 | |
| AL832866.1 | 4 | 2826 | CAI46196.1 | |
| BC000880.1 | 5 | 803 | AAH00880.1 | |
| BC015095.1 | 5 | 812 | ||
| BC095446.1 | 4 | 804 | AAH95446.1 | |
| BM806646.1 | 6 | 941 | ||
| CB158821.1 | 3 | 531 |
More Information
Set Membership: |
|

Genomic Map