Assay Details
Target Gene Details
Entrez Gene ID: | 9156 |
Gene Name: | exonuclease 1 |
Gene Aliases: |
HEX1, hExoI |
Location: |
Chr.1:241847967-241889939 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXO1 | NM_001319224.1 | 2 | 262 | NP_001306153.1 |
| NM_003686.4 | 1 | 99 | NP_003677.4 | |
| NM_006027.4 | 1 | 99 | NP_006018.4 | |
| NM_130398.3 | 2 | 262 | NP_569082.2 | |
| XM_006711840.2 | 2 | 262 | XP_006711903.1 | |
| XM_011544321.1 | 2 | 264 | XP_011542623.1 | |
| XM_011544322.1 | 2 | 333 | XP_011542624.1 | |
| XM_011544323.2 | 2 | 262 | XP_011542625.1 | |
| XM_011544324.2 | 2 | 262 | XP_011542626.1 | |
| XM_017002793.1 | 2 | 264 | XP_016858282.1 | |
| AF042282.1 | AAC32259.1 | |||
| AF060479.1 | AAC33874.1 | |||
| AF084974.1 | AAD13754.1 | |||
| AF091740.1 | AAC63043.1 | |||
| AK291291.1 | 2 | 250 | ||
| BC007491.2 | 2 | 210 | AAH07491.1 | |
| CD644038.1 | 2 | 230 | ||
| CN348533.1 | 2 | 247 | ||
| DA479425.1 | 2 | 265 | ||
| DA661516.1 | 2 | 263 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762521 | Chr.1:241727738 - 241910920 on Build GRCh38 | Loss |
|
| nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain |
|
| nsv528015 | Chr.1:241701917 - 241907429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map