Assay Details
Target Gene Details
Entrez Gene ID: | 6311 |
Gene Name: | ataxin 2 |
Gene Aliases: |
ASL13, ATX2, SCA2, TNRC13 |
Location: |
Chr.12:111452214-111599676 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATXN2 | NM_001310121.1 | 26 | 3826 | NP_001297050.1 |
| NM_001310123.1 | 22 | 3588 | NP_001297052.1 | |
| NM_002973.3 | 25 | 4545 | NP_002964.3 | |
| NR_132311.1 | 24 | 4484 | ||
| AA731027.1 | 1 | 168 | ||
| AI083696.1 | 1 | 159 | ||
| AI668891.1 | 1 | 158 | ||
| AL042619.2 | 1 | 467 | ||
| BF448025.1 | 1 | 120 | ||
| BM671308.1 | 1 | 175 | ||
| Y08262.1 | 24 | 4081 | CAA69589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832513 | Chr.12:111284287 - 111455130 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map