Assay Details
Target Gene Details
Entrez Gene ID: | 51125 |
Gene Name: | golgin A7 |
Gene Aliases: |
GCP16, GOLGA3AP1, GOLGA7A, HSPC041 |
Location: |
Chr.8:41490562-41510980 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA7 | NM_001002296.1 | 1 | 210 | NP_001002296.1 |
| NM_001174124.1 | NP_001167595.1 | |||
| NM_016099.2 | NP_057183.2 | |||
| AB104615.1 | BAC82368.1 | |||
| AF068291.1 | AAF65180.1 | |||
| AF125102.1 | AAD39919.1 | |||
| AK026613.1 | ||||
| AL695304.1 | ||||
| BC012032.1 | 1 | 210 | AAH12032.1 | |
| BI553491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831299 | Chr.8:41448617 - 41692486 on Build GRCh38 | Loss |
|
| esv3576565 | Chr.8:41064045 - 41790840 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map