Assay Details
Target Gene Details
Entrez Gene ID: | 7786 |
Gene Name: | mitogen-activated protein kinase kinase kinase 12 |
Gene Aliases: |
DLK, MEKK12, MUK, ZPK, ZPKP1 |
Location: |
Chr.12:53480492-53501066 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K12 | NM_001193511.1 | 14 | 3188 | NP_001180440.1 |
| NM_006301.3 | 15 | 3089 | NP_006292.3 | |
| XM_005269138.3 | 14 | 3116 | XP_005269195.1 | |
| XM_006719588.3 | 14 | 3010 | XP_006719651.1 | |
| XM_011538725.2 | 14 | 3993 | XP_011537027.1 | |
| XM_017019956.1 | 15 | 3017 | XP_016875445.1 | |
| AB209453.1 | 5 | 5173 | BAD92690.1 | |
| BC050050.1 | 11 | 2069 | AAH50050.1 | |
| U07358.1 | 15 | 2916 | AAA67343.1 |
Target Gene Details
Entrez Gene ID: | 5094 |
Gene Name: | poly(rC) binding protein 2 |
Gene Aliases: |
HNRNPE2, HNRPE2, hnRNP-E2 |
Location: |
Chr.12:53452102-53481162 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBP2 | NM_001098620.2 | 14 | 2848 | NP_001092090.1 |
| NM_001128911.1 | 15 | 2950 | NP_001122383.1 | |
| NM_001128912.1 | 15 | 2938 | NP_001122384.1 | |
| NM_001128913.1 | 14 | 2860 | NP_001122385.1 | |
| NM_001128914.1 | 13 | 2809 | NP_001122386.1 | |
| NM_005016.5 | 15 | 2953 | NP_005007.2 | |
| NM_031989.4 | 15 | 2941 | NP_114366.1 | |
| BF509278.1 | 1 | 235 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv714 | Chr.12:53455169 - 53489159 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map