Assay Details
Target Gene Details
Entrez Gene ID: | 56935 |
Gene Name: | single-pass membrane protein with coiled-coil domains 4 |
Gene Aliases: |
C11orf75, FN5 |
Location: |
Chr.11:93478472-93586856 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMCO4 | NM_020179.2 | 3 | 570 | NP_064564.1 |
| XM_011542907.1 | 3 | 595 | XP_011541209.1 | |
| XM_011542908.2 | 4 | 698 | XP_011541210.1 | |
| XM_011542909.1 | 3 | 580 | XP_011541211.1 | |
| XM_011542910.2 | 3 | 1333 | XP_011541212.1 | |
| XM_011542911.2 | 3 | 1259 | XP_011541213.1 | |
| XM_017018019.1 | 3 | 1315 | XP_016873508.1 | |
| XM_017018020.1 | 4 | 756 | XP_016873509.1 | |
| AF197137.1 | 3 | 542 | AAF86046.1 | |
| BC031564.1 | 3 | 570 | AAH31564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760208 | Chr.11:93439507 - 93533428 on Build GRCh38 | Loss |
|
| nsv1039134 | Chr.11:93056834 - 93486776 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map