Assay Details
Target Gene Details
Entrez Gene ID: | 51699 |
Gene Name: | VPS29, retromer complex component |
Gene Aliases: |
DC15, DC7, PEP11 |
Location: |
Chr.12:110491523-110502140 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS29 | NM_001282150.1 | 6 | 755 | NP_001269079.1 |
| NM_001282151.1 | 3 | 364 | NP_001269080.1 | |
| NM_016226.4 | 4 | 600 | NP_057310.1 | |
| NM_057180.2 | 5 | 612 | NP_476528.1 | |
| NR_104099.1 | 6 | 652 | ||
| NR_104100.1 | 5 | 640 | ||
| NR_104101.1 | 5 | 690 | ||
| NR_104102.1 | 4 | 491 | ||
| XM_006719459.3 | 6 | 661 | XP_006719522.1 | |
| XM_006719460.3 | 7 | 698 | XP_006719523.1 | |
| XM_011538484.2 | 5 | 646 | XP_011536786.1 | |
| XM_017019457.1 | 5 | 671 | XP_016874946.1 | |
| XM_017019458.1 | 4 | 791 | XP_016874947.1 | |
| XM_017019459.1 | 4 | 453 | XP_016874948.1 | |
| AA148059.1 | 4 | 437 | ||
| AB578890.1 | ||||
| AF168716.1 | 5 | 570 | AAF87318.1 | |
| AF175264.1 | 4 | 569 | AAF89952.1 | |
| AF193795.1 | 4 | 562 | AAF04596.1 | |
| AF201936.1 | 5 | 657 | AAF86872.1 | |
| AF201946.1 | 4 | 548 | AAF17238.1 | |
| AK000499.1 | 3 | 328 | ||
| AK129767.1 | 4 | 562 | ||
| AL136614.1 | 4 | 578 | CAB66549.1 | |
| AL832866.1 | 4 | 2593 | CAI46196.1 | |
| BC000880.1 | 5 | 570 | AAH00880.1 | |
| BC015095.1 | 5 | 579 | ||
| BC095446.1 | 4 | 571 | AAH95446.1 | |
| BG707325.1 | 4 | 595 | ||
| BM806646.1 | 6 | 708 | ||
| BU568689.1 | 6 | 613 | ||
| CB158821.1 | 3 | 298 | ||
| CR457182.1 | CAG33463.1 | |||
| CR533468.1 | CAG38499.1 |
More Information
Set Membership: |
|

Genomic Map