Assay Details
Target Gene Details
Entrez Gene ID: | 27030 |
Gene Name: | mutL homolog 3 |
Gene Aliases: |
HNPCC7 |
Location: |
Chr.14:75013764-75051532 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLH3 | NM_001040108.1 | 13 | 4736 | NP_001035197.1 |
| NM_014381.2 | 12 | 4664 | NP_055196.2 | |
| XM_005267532.4 | 12 | 4631 | XP_005267589.1 | |
| XM_005267533.4 | 11 | 4559 | XP_005267590.1 | |
| XM_006720116.3 | 13 | 4648 | XP_006720179.1 | |
| XM_011536646.2 | 11 | 4565 | XP_011534948.1 | |
| XM_017021219.1 | 12 | 4590 | XP_016876708.1 | |
| AF195657.1 | 12 | 4562 | AAF23904.1 | |
| AL833875.1 | 12 | 2566 | CAD38732.1 | |
| BC036866.2 | 10 | 1168 | AAH36866.1 | |
| L40399.1 | 12 | 1406 | AAC42005.1 |
More Information
Set Membership: |
|

Genomic Map