Assay Details
Target Gene Details
Entrez Gene ID: | 285761 |
Gene Name: | discoidin, CUB and LCCL domain containing 1 |
Gene Aliases: |
dJ94G16.1 |
Location: |
Chr.6:117480346-117569858 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCBLD1 | NM_173674.2 | NP_775945.1 | ||
| BC035671.1 | AAH35671.1 | |||
| CR749448.1 | CAH18285.1 |
Target Gene Details
Entrez Gene ID: | 57120 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
CAL, FIG, GOPC1, PIST, dJ94G16.2 |
Location: |
Chr.6:117560269-117602542 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOPC | NM_001017408.2 | 8 | 4235 | NP_001017408.1 |
| NM_020399.3 | 9 | 4259 | NP_065132.1 | |
| AB209385.1 | 8 | 5328 | BAD92622.1 | |
| AJ420540.1 | 1 | 2231 | ||
| AK025074.1 | 1 | 2120 | ||
| AY033606.1 | 8 | 4188 | AAK57733.1 |
More Information
Set Membership: |
|

Genomic Map