Assay Details
Target Gene Details
Entrez Gene ID: | 548645 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C25 |
Gene Aliases: |
bA16L21.2.1 |
Location: |
Chr.9:111631352-111654351 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC25 | NM_001015882.2 | 4 | 1813 | NP_001015882.2 |
| NR_037148.1 | 5 | 2051 | ||
| AF447177.1 | 5 | 2020 | AAQ04643.1 | |
| AK095452.1 | 5 | 2034 | ||
| AW629467.1 | 1 | 466 |
Target Gene Details
Entrez Gene ID: | 552891 |
Gene Name: | DNAJC25-GNG10 readthrough |
Gene Aliases: |
- |
Location: |
Chr.9:111631352-111670246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC25-GNG10 | NM_004125.3 | NP_004116.2 | ||
| BC015391.1 | AAH15391.1 | |||
| BT007068.1 | AAP35731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825041 | Chr.9:111607177 - 111673958 on Build GRCh38 | Gain |
|
| nsv615192 | Chr.9:111624319 - 111661153 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map