Assay Details
Target Gene Details
Entrez Gene ID: | 7776 |
Gene Name: | zinc finger protein 236 |
Gene Aliases: |
ZNF236A, ZNF236B |
Location: |
Chr.18:76822550-76970732 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF236 | NM_001306089.1 | 2 | 98 | NP_001293018.1 |
| NM_007345.3 | 2 | 289 | NP_031371.3 | |
| XM_005266762.3 | 2 | 441 | XP_005266819.1 | |
| XM_011526165.2 | 3 | 443 | XP_011524467.1 | |
| XM_011526168.2 | 2 | 155 | XP_011524470.1 | |
| XM_011526169.2 | 2 | 154 | XP_011524471.1 | |
| AF085243.1 | 2 | 289 | AAD55328.1 | |
| AF085244.1 | 2 | 289 | AAD55329.1 | |
| BC140859.1 | 2 | 120 | ||
| BX648028.1 | 2 | 98 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
|
| nsv1056690 | Chr.18:76794730 - 76903712 on Build GRCh38 | Gain |
|
| nsv1063632 | Chr.18:76317786 - 77170410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map