Assay Details
Target Gene Details
Entrez Gene ID: | 23035 |
Gene Name: | PH domain and leucine rich repeat protein phosphatase 2 |
Gene Aliases: |
PHLPPL, PPM3B |
Location: |
Chr.16:71644924-71724701 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLPP2 | NM_001289003.1 | 18 | 4350 | NP_001275932.1 |
| NM_015020.3 | 19 | 4204 | NP_055835.2 | |
| AB023148.2 | 18 | 4350 | ||
| AK001661.1 | 1 | 733 | ||
| AK001854.1 | BAA91943.1 | |||
| BC035267.1 | AAH35267.1 | |||
| BC129927.1 | 18 | 3831 | ||
| BC129983.1 | ||||
| BX647823.1 | 19 | 4205 | ||
| BX648157.1 | 13 | 2940 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv962218 | Chr.16:71611299 - 71693703 on Build GRCh38 | Duplication |
|
| nsv1063338 | Chr.16:71578443 - 71719065 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map