Assay Details
Target Gene Details
Entrez Gene ID: | 285172 |
Gene Name: | family with sequence similarity 126 member B |
Gene Aliases: |
HYCC2 |
Location: |
Chr.2:200973718-201071671 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM126B | NM_001321618.1 | 11 | 2225 | NP_001308547.1 |
| NM_001321619.1 | 12 | 2436 | NP_001308548.1 | |
| NM_001321621.1 | 11 | 2338 | NP_001308550.1 | |
| NM_001321622.1 | 13 | 2554 | NP_001308551.1 | |
| NM_001321623.1 | 13 | 2491 | NP_001308552.1 | |
| NM_001321624.1 | 12 | 2393 | NP_001308553.1 | |
| NM_001321625.1 | 13 | 2604 | NP_001308554.1 | |
| NM_001321626.1 | 13 | 2624 | NP_001308555.1 | |
| NM_001321627.1 | 14 | 2547 | NP_001308556.1 | |
| NM_001321628.1 | 14 | 2660 | NP_001308557.1 | |
| NM_001321629.1 | 13 | 2379 | NP_001308558.1 | |
| NM_173822.4 | 12 | 2323 | NP_776183.1 | |
| XM_006712443.3 | 12 | 2506 | XP_006712506.1 | |
| XM_011511002.2 | 13 | 2521 | XP_011509304.1 | |
| XM_017003879.1 | 14 | 2702 | XP_016859368.1 | |
| XM_017003880.1 | 13 | 3116 | XP_016859369.1 | |
| XM_017003881.1 | 12 | 2384 | XP_016859370.1 | |
| XM_017003882.1 | 14 | 2603 | XP_016859371.1 | |
| XM_017003883.1 | 13 | 2778 | XP_016859372.1 | |
| AK027168.1 | 1 | 198 | ||
| AK097111.1 | 5 | 1566 | ||
| AL832234.1 | 11 | 2182 | ||
| AL832346.1 | 14 | 2542 | ||
| AL833403.1 | 12 | 2307 | ||
| BC039295.1 | 12 | 2321 | AAH39295.1 |
More Information
Set Membership: |
|

Genomic Map