Assay Details
Target Gene Details
Entrez Gene ID: | 10992 |
Gene Name: | splicing factor 3b subunit 2 |
Gene Aliases: |
Cus1, SAP145, SF3B145, SF3b1, SF3b150 |
Location: |
Chr.11:66051772-66068911 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B2 | NM_006842.2 | 1 | 73 | NP_006833.2 |
| XM_005273726.3 | 1 | 646 | XP_005273783.1 | |
| XM_011544740.2 | 1 | 646 | XP_011543042.1 | |
| XM_017017144.1 | 1 | 646 | XP_016872633.1 | |
| AK290850.1 | 1 | 57 | ||
| BC000401.2 | AAH00401.2 | |||
| BC053577.1 | 1 | 40 | AAH53577.1 | |
| BX537771.1 | 1 | 40 | CAD97834.1 | |
| U41371.1 | AAA97461.1 |
More Information
Set Membership: |
|

Genomic Map