Assay Details
Target Gene Details
Entrez Gene ID: | 57149 |
Gene Name: | LYR motif containing 1 |
Gene Aliases: |
A211C6.1 |
Location: |
Chr.16:20899626-20925006 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYRM1 | NM_001128301.2 | 5 | 1565 | NP_001121773.1 |
| NM_001128302.2 | 4 | 1333 | NP_001121774.1 | |
| NM_001302835.1 | 6 | 1509 | NP_001289764.1 | |
| NM_001302836.1 | 6 | 1661 | NP_001289765.1 | |
| NM_020424.4 | 5 | 1484 | NP_065157.1 | |
| NR_126516.1 | 4 | 1406 | ||
| NR_126520.1 | 3 | 1188 | ||
| NR_126521.1 | 3 | 1174 | ||
| NR_126527.1 | 3 | 1002 | ||
| NR_126528.1 | 4 | 1420 | ||
| XM_005255444.1 | 5 | 1386 | XP_005255501.1 | |
| XM_011545900.2 | 5 | 1148 | XP_011544202.1 | |
| XM_011545901.2 | 5 | 1214 | XP_011544203.1 | |
| XM_011545902.1 | 6 | 1485 | XP_011544204.1 | |
| XM_011545908.2 | 5 | 1216 | XP_011544210.1 | |
| XM_017023475.1 | 6 | 1901 | XP_016878964.1 | |
| XM_017023476.1 | 4 | 1799 | XP_016878965.1 | |
| XM_017023477.1 | 4 | 1736 | XP_016878966.1 | |
| XM_017023478.1 | 5 | 1206 | XP_016878967.1 | |
| XM_017023479.1 | 5 | 1643 | XP_016878968.1 | |
| XM_017023480.1 | 7 | 1729 | XP_016878969.1 | |
| XM_017023481.1 | 5 | 1358 | XP_016878970.1 | |
| XM_017023482.1 | 4 | 1412 | XP_016878971.1 | |
| AK308690.1 | 3 | 1253 | ||
| AL359616.1 | 1 | 586 | ||
| BC017039.1 | 6 | 1509 | AAH17039.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457444 | Chr.16:20917903 - 21160552 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|

Genomic Map