Assay Details
Target Gene Details
Entrez Gene ID: | 9616 |
Gene Name: | ring finger protein 7 |
Gene Aliases: |
CKBBP1, ROC2, SAG |
Location: |
Chr.3:141738209-141746803 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF7 | NM_001201370.1 | 2 | 781 | NP_001188299.1 |
| NM_014245.4 | 3 | 829 | NP_055060.1 | |
| NM_183237.2 | 3 | 822 | NP_899060.1 | |
| NR_037702.1 | 4 | 1093 | ||
| NR_037703.1 | 4 | 1086 | ||
| AF092878.1 | AAD25962.1 | |||
| AF164679.1 | AAD55984.1 | |||
| BC005966.1 | AAH05966.1 | |||
| BC008627.1 | AAH08627.1 | |||
| BF439211.1 | ||||
| BG716023.1 | ||||
| BG742338.1 |
More Information
Set Membership: |
|

Genomic Map