Assay Details
Target Gene Details
Entrez Gene ID: | 163404 |
Gene Name: | phospholipid phosphatase related 5 |
Gene Aliases: |
LPPR5, PAP2, PAP2D, PRG5 |
Location: |
Chr.1:98890245-99005850 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPPR5 | NM_001010861.2 | 6 | 3557 | NP_001010861.1 |
| NM_001037317.1 | 6 | 3572 | NP_001032394.1 | |
| XM_011540836.2 | 7 | 3629 | XP_011539138.1 | |
| XM_011540838.2 | 7 | 4047 | XP_011539140.1 | |
| AF131783.1 | 1 | 1084 | ||
| BC040174.1 | 6 | 3294 | AAH40174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830771 | Chr.1:98877965 - 99049970 on Build GRCh38 | Loss |
|
| nsv830760 | Chr.1:98730523 - 98898996 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map