Assay Details
Target Gene Details
Entrez Gene ID: | 7090 |
Gene Name: | transducin like enhancer of split 3 |
Gene Aliases: |
ESG, ESG3, GRG3, HsT18976 |
Location: |
Chr.15:70047790-70097935 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951987 | Chr.15:70043962 - 70061561 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map