Assay Details
Target Gene Details
Entrez Gene ID: | 27097 |
Gene Name: | TATA-box binding protein associated factor 5 like |
Gene Aliases: |
PAF65B |
Location: |
Chr.1:229593111-229626047 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF5L | NM_001025247.1 | 4 | 739 | NP_001020418.1 |
| NM_014409.3 | 4 | 739 | NP_055224.1 | |
| XM_005273099.4 | 4 | 692 | XP_005273156.1 | |
| XM_005273100.2 | 5 | 705 | XP_005273157.1 | |
| XM_011544162.1 | 3 | 476 | XP_011542464.1 | |
| XM_011544163.1 | 7 | 1321 | XP_011542465.1 | |
| XM_011544164.2 | 3 | 419 | XP_011542466.1 | |
| XM_017001045.1 | 6 | 1476 | XP_016856534.1 | |
| AF069736.1 | 3 | 573 | AAC39906.1 | |
| BC041094.1 | 4 | 662 | AAH41094.1 |
More Information
Set Membership: |
|

Genomic Map