Assay Details
Target Gene Details
Entrez Gene ID: | 5162 |
Gene Name: | pyruvate dehydrogenase (lipoamide) beta |
Gene Aliases: |
PDHBD, PDHE1-B, PHE1B |
Location: |
Chr.3:58427630-58433852 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDHB | NM_000925.3 | NP_000916.2 | ||
| NM_001173468.1 | NP_001166939.1 | |||
| NM_001315536.1 | NP_001302465.1 | |||
| NR_033384.1 | ||||
| AB209241.1 | 2 | 1707 | BAD92478.1 | |
| AK225569.1 | ||||
| AK225621.1 | ||||
| AK293153.1 | ||||
| AK311514.1 | ||||
| AK313022.1 | ||||
| AL117618.1 | CAB56017.1 | |||
| BC000439.2 | AAH00439.1 | |||
| BC001924.1 | AAH01924.1 | |||
| CR541911.1 | CAG46709.1 | |||
| DA965753.1 | ||||
| DB039833.1 | ||||
| J03576.1 | AAA88097.1 | |||
| M19123.1 | AAA60052.1 | |||
| M34055.1 | AAA60233.1 | |||
| M34056.1 | AAA60054.1 | |||
| M34479.1 | AAA36428.1 | |||
| M54788.1 | AAA60053.1 | |||
| X57778.1 | CAA40924.1 |
More Information
Set Membership: |
|

Genomic Map