Assay Details
Target Gene Details
Entrez Gene ID: | 163351 |
Gene Name: | guanylate binding protein family member 6 |
Gene Aliases: |
- |
Location: |
Chr.1:89363877-89388160 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP6 | NM_001320257.1 | 9 | 2862 | NP_001307186.1 |
| NM_198460.2 | 11 | 3203 | NP_940862.2 | |
| XM_011540835.2 | 11 | 3032 | XP_011539137.1 | |
| AI199897.1 | ||||
| AK096454.1 | 1 | 453 | ||
| BX537949.1 | CAD97917.1 |
More Information
Set Membership: |
|

Genomic Map