Assay Details
Target Gene Details
Entrez Gene ID: | 4110 |
Gene Name: | MAGE family member A11 |
Gene Aliases: |
CT1.11, MAGE-11, MAGE11, MAGEA-11 |
Location: |
Chr.X:149688202-149717268 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA11 | NM_001011544.1 | NP_001011544.1 | ||
| NM_005366.4 | NP_005357.2 | |||
| XM_017029521.1 | XP_016885010.1 | |||
| XM_017029522.1 | XP_016885011.1 | |||
| AK127849.1 | BAC87161.1 | |||
| AY747607.1 | AAW71787.1 | |||
| BC004479.2 | AAH04479.2 | |||
| BQ212898.1 | ||||
| BX337921.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525908 | Chr.X:148833079 - 150525125 on Build GRCh38 | Gain |
|
| esv3577515 | Chr.X:149619941 - 149746113 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map