Assay Details
Target Gene Details
Entrez Gene ID: | 219960 |
Gene Name: | olfactory receptor family 10 subfamily Q member 1 |
Gene Aliases: |
OR11-233 |
Location: |
Chr.11:58227882-58228918 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10Q1 | NM_001004471.2 | 1 | 374 | NP_001004471.1 |
| BC140731.1 | 1 | 374 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832166 | Chr.11:58084559 - 58238717 on Build GRCh38 | Gain+Loss |
|
| nsv832165 | Chr.11:58038552 - 58241604 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map