Assay Details
Target Gene Details
Entrez Gene ID: | 51438 |
Gene Name: | MAGE family member C2 |
Gene Aliases: |
CT10, HCA587, MAGEE1 |
Location: |
Chr.X:142202342-142205290 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC2 | NM_016249.3 | 3 | 1898 | NP_057333.1 |
| AF116194.1 | 3 | 1881 | AAF34816.1 | |
| AF151378.1 | 3 | 1879 | AAF36533.1 | |
| AF196482.1 | 3 | 1879 | AAF07210.1 | |
| BC005891.2 | 1 | 1629 | AAH05891.1 | |
| BC013318.1 | 3 | 1902 | AAH13318.1 | |
| BM548987.1 | 1 | 354 |
More Information
Set Membership: |
|

Genomic Map