Assay Details
Target Gene Details
Entrez Gene ID: | 79632 |
Gene Name: | family with sequence similarity 184 member A |
Gene Aliases: |
C6orf60 |
Location: |
Chr.6:118959829-119149193 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM184A | NM_001100411.2 | 17 | 3289 | NP_001093881.1 |
| NM_001288576.1 | 16 | 3184 | NP_001275505.1 | |
| NM_024581.5 | 18 | 3827 | NP_078857.5 | |
| AJ420503.1 | 2 | 2307 | ||
| AJ420563.1 | 1 | 1214 | ||
| AK024004.1 | 11 | 1631 | BAB14769.1 | |
| AK091682.1 | 16 | 3180 | BAC03719.1 | |
| AK127744.1 | 14 | 2129 | BAC87110.1 | |
| AL136767.1 | 8 | 1037 | CAB66701.1 | |
| AU160036.1 | 1 | 219 | ||
| AY131226.1 | 14 | 2103 | AAN07912.1 | |
| BC009055.1 | 10 | 1419 | AAH09055.2 | |
| BC060769.1 | 18 | 3827 | AAH60769.2 | |
| BX538186.1 | 17 | 3269 | CAD98058.1 |
More Information
Set Membership: |
|

Genomic Map