Assay Details
Target Gene Details
Entrez Gene ID: | 402176 |
Gene Name: | ribosomal protein L21 pseudogene 44 |
Gene Aliases: |
RPL21_17_477 |
Location: |
Chr.4:53985486-53987282 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL21P44 | NR_027153.1 | 1 | 966 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv966269 | Chr.4:53985914 - 53987301 on Build GRCh38 | Duplication |
|

Genomic Map