Assay Details
Target Gene Details
Entrez Gene ID: | 1553 |
Gene Name: | cytochrome P450 family 2 subfamily A member 13 |
Gene Aliases: |
CPAD, CYP2A, CYPIIA13 |
Location: |
Chr.19:41088451-41096195 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2A13 | NM_000766.4 | NP_000757.2 | ||
| AF209774.1 | AAG35775.1 | |||
| AY513604.1 | AAR90935.1 | |||
| AY513605.1 | AAR90936.1 | |||
| AY513606.1 | AAR90937.1 | |||
| AY513607.1 | AAR90938.1 | |||
| AY513608.1 | AAR90939.1 | |||
| AY513609.1 | AAR90940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2491 | Chr.19:41055043 - 41098317 on Build GRCh38 | Deletion |
|
| nsv978819 | Chr.19:41083984 - 41095410 on Build GRCh38 | Duplication |
|
| nsv1065684 | Chr.19:41010484 - 41138637 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map