Assay Details
Target Gene Details
Entrez Gene ID: | 219983 |
Gene Name: | olfactory receptor family 4 subfamily D member 6 |
Gene Aliases: |
OR11-250 |
Location: |
Chr.11:59456961-59457905 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D6 | NM_001004708.1 | NP_001004708.1 | ||
| BC137028.1 | 1 | 965 | ||
| BC137029.1 | 1 | 965 |
More Information
Additional Information:
For this assay, SNP(s) [rs116863593] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map