Assay Details
Target Gene Details
Entrez Gene ID: | 100533464 |
Gene Name: | PRH1-PRR4 readthrough |
Gene Aliases: |
PRH1 |
Location: |
Chr.12:10845849-11171625 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1-PRR4 | NR_037918.2 | |||
| AB593030.1 |
Target Gene Details
Entrez Gene ID: | 11272 |
Gene Name: | proline rich 4 (lacrimal) |
Gene Aliases: |
LPRP, PROL4 |
Location: |
Chr.12:10845849-10849476 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR4 | NM_001098538.2 | NP_001092008.2 | ||
| NM_007244.2 | NP_009175.2 | |||
| AF530472.1 | AAM94338.1 | |||
| AK292934.1 | ||||
| AK292991.1 | ||||
| AK304321.1 | ||||
| BC058035.1 | AAH58035.1 | |||
| BX537839.1 | ||||
| DB229285.1 | ||||
| S79048.1 | AAB35174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628543 | Chr.12:10720386 - 10881212 on Build GRCh38 | Gain |
|
| esv2759879 | Chr.12:10817021 - 11561987 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map