Assay Details
Target Gene Details
Entrez Gene ID: | 55764 |
Gene Name: | intraflagellar transport 122 |
Gene Aliases: |
CED, CED1, SPG, WDR10, WDR10p, WDR140 |
Location: |
Chr.3:129440036-129520507 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2725901 | Chr.3:129380821 - 129487939 on Build GRCh38 | Deletion |
|
| esv3597741 | Chr.3:129383049 - 129486193 on Build GRCh38 | Loss |
|
| nsv591668 | Chr.3:129403449 - 129490335 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map