Assay Details
Target Gene Details
Entrez Gene ID: | 340441 |
Gene Name: | POTE ankyrin domain family member A |
Gene Aliases: |
A26A1, CT104.3, POTE-8, POTE8 |
Location: |
Chr.8:43292370-43363185 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POTEA | NM_001002920.1 | NP_001002920.1 | ||
| NM_001005365.2 | 5 | 824 | NP_001005365.2 | |
| XM_011544516.1 | 5 | 896 | XP_011542818.1 | |
| AY462869.1 | 5 | 824 | AAS58863.1 | |
| AY462870.1 | AAS58864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv981932 | Chr.8:43292470 - 43338145 on Build GRCh38 | Duplication |
|
| dgv7187n100 | Chr.8:43081800 - 43968893 on Build GRCh38 | Gain |
|
| esv3576566 | Chr.8:43192859 - 43931580 on Build GRCh38 | Gain |
|
| esv3891387 | Chr.8:42998571 - 43918067 on Build GRCh38 | Gain |
|
| nsv470208 | Chr.8:43082465 - 43936548 on Build GRCh38 | Gain |
|
| nsv465644 | Chr.8:43231785 - 43431234 on Build GRCh38 | Gain |
|
| dgv12123n54 | Chr.8:43169482 - 43937428 on Build GRCh38 | Gain |
|
| nsv1032781 | Chr.8:42917251 - 43606341 on Build GRCh38 | Gain |
|
| dgv7188n100 | Chr.8:43152014 - 43726076 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map