Assay Details
Target Gene Details
Entrez Gene ID: | 2635 |
Gene Name: | guanylate binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.1:89006677-89022880 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP3 | NM_001319179.1 | 12 | 3007 | NP_001306108.1 |
| NM_001319180.1 | 11 | 3063 | NP_001306109.1 | |
| NM_018284.2 | 11 | 2921 | NP_060754.2 | |
| NR_135007.1 | 9 | 2427 | ||
| XM_005270743.2 | 12 | 2997 | XP_005270800.1 | |
| XM_006710574.2 | 11 | 2854 | XP_006710637.1 | |
| XM_011541233.1 | 10 | 2709 | XP_011539535.1 | |
| XM_011541234.1 | 10 | 2692 | XP_011539536.1 | |
| XM_017001013.1 | 10 | 2841 | XP_016856502.1 | |
| XM_017001014.1 | 9 | 2589 | XP_016856503.1 | |
| AK309096.1 | 10 | 3561 | ||
| AL136680.1 | 11 | 2820 | CAB66615.1 | |
| BU678920.1 | 1 | 131 | ||
| CA450307.1 | 1 | 131 | ||
| CR936755.1 | 11 | 2922 | ||
| CR936758.1 | 11 | 2862 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain |
|
| nsv546785 | Chr.1:88994245 - 89010744 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map