Assay Details
Target Gene Details
Entrez Gene ID: | 64431 |
Gene Name: | ARP6 actin-related protein 6 homolog |
Gene Aliases: |
ARP6, CDA12, HSPC281, MSTP136, hARP6, hARPX |
Location: |
Chr.12:100200087-100224425 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR6 | NM_022496.4 | 11 | 1892 | NP_071941.1 |
| NR_048568.1 | 11 | 1343 | ||
| NR_048569.1 | 11 | 1215 | ||
| XM_017019821.1 | 11 | 2046 | XP_016875310.1 | |
| AB038229.1 | 11 | 1177 | BAB20762.1 | |
| AF175226.1 | 11 | 1133 | AAQ13651.1 | |
| AF212251.1 | 11 | 1170 | AAK14934.1 | |
| AK023495.1 | 11 | 1162 | BAB14588.1 | |
| AK023684.1 | 11 | 1176 | BAB14640.1 | |
| AK124075.1 | 11 | 1892 | ||
| AK222959.1 | 11 | 1171 | BAD96679.1 | |
| BC015107.1 | 11 | 1187 | AAH15107.1 |
Target Gene Details
Entrez Gene ID: | 120863 |
Gene Name: | DEP domain containing 4 |
Gene Aliases: |
DEP.4 |
Location: |
Chr.12:100203669-100268094 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEPDC4 | NR_135073.1 | |||
| AK090824.1 | BAC03525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892256 | Chr.12:100007917 - 100225312 on Build GRCh38 | Loss |
|
| nsv1048784 | Chr.12:100058759 - 100251225 on Build GRCh38 | Gain |
|
| esv3630550 | Chr.12:100215678 - 100242288 on Build GRCh38 | Gain |
|
| nsv559964 | Chr.12:99872025 - 100538490 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map