Assay Details
Target Gene Details
Entrez Gene ID: | 284217 |
Gene Name: | laminin subunit alpha 1 |
Gene Aliases: |
LAMA, PTBHS, S-LAM-alpha |
Location: |
Chr.18:6941744-7117814 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 57 - Exon 58 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMA1 | NM_005559.3 | NP_005550.2 | ||
| XM_011525655.2 | XP_011523957.1 | |||
| XM_011525656.2 | XP_011523958.1 | |||
| AF351616.1 | 3 | 2873 | AAN76516.1 | |
| AK091949.1 | ||||
| AK093376.1 | 1 | 1013 | ||
| BC039051.1 | AAH39051.1 | |||
| BC062318.1 | AAH62318.1 |
Target Gene Details
Entrez Gene ID: | 101927188 |
Gene Name: | uncharacterized LOC101927188 |
Gene Aliases: |
- |
Location: |
Chr.18:6954677-6957419 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927188 | NR_126040.1 | 2 | 311 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833583 | Chr.18:6863009 - 7011757 on Build GRCh38 | Gain |
|
| nsv576397 | Chr.18:6758922 - 7036684 on Build GRCh38 | Gain |
|
| nsv1064175 | Chr.18:6740268 - 7384530 on Build GRCh38 | Gain |
|
| esv3584943 | Chr.18:6873355 - 7434331 on Build GRCh38 | Gain |
|
| esv3641677 | Chr.18:6930548 - 7042397 on Build GRCh38 | Gain |
|
| nsv1064849 | Chr.18:6936252 - 8070494 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map