Assay Details
Target Gene Details
Entrez Gene ID: | 55758 |
Gene Name: | REST corepressor 3 |
Gene Aliases: |
- |
Location: |
Chr.1:211259347-211316385 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
More Information
Set Membership: |
|

Genomic Map