Assay Details
Target Gene Details
Entrez Gene ID: | 51103 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 1 |
Gene Aliases: |
CGI-65, CGI65, CIA30 |
Location: |
Chr.15:41387349-41402694 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF1 | NM_016013.3 | 2 | 809 | NP_057097.2 |
| NR_045620.1 | 2 | 809 | ||
| XM_006720555.2 | 2 | 739 | XP_006720618.1 | |
| XM_011521658.1 | 2 | 673 | XP_011519960.1 | |
| XM_011521659.2 | 2 | 836 | XP_011519961.1 | |
| XM_017022293.1 | 2 | 740 | XP_016877782.1 | |
| AF151823.1 | 2 | 771 | AAD34060.1 | |
| AK309889.1 | 2 | 809 | ||
| AK310983.1 | 2 | 809 | ||
| BC000780.2 | 2 | 793 | AAH00780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039283 | Chr.15:41328301 - 41453282 on Build GRCh38 | Gain |
|
| dgv2580n100 | Chr.15:41267962 - 41453282 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map