Assay Details
Target Gene Details
Entrez Gene ID: | 9452 |
Gene Name: | integral membrane protein 2A |
Gene Aliases: |
BRICD2A, E25A |
Location: |
Chr.X:79360384-79367552 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITM2A | NM_001171581.1 | 3 | 701 | NP_001165052.1 |
| NM_004867.4 | 4 | 833 | NP_004858.1 | |
| AB209310.1 | 1 | 4660 | BAD92547.1 | |
| AF038953.1 | 4 | 635 | AAC39867.1 | |
| AK223327.1 | 4 | 612 | BAD97047.1 | |
| AK303238.1 | 3 | 483 | ||
| AK313154.1 | 4 | 791 | ||
| AY359051.1 | 4 | 574 | AAQ89410.1 | |
| BC040437.1 | 4 | 588 | AAH40437.1 | |
| BM561934.1 | 1 | 76 | ||
| CR456875.1 | 4 | 496 | CAG33156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519126 | Chr.X:79331247 - 79373681 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map