Assay Details
Target Gene Details
Entrez Gene ID: | 10256 |
Gene Name: | connector enhancer of kinase suppressor of Ras 1 |
Gene Aliases: |
CNK, CNK1 |
Location: |
Chr.1:26177490-26189884 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNKSR1 | NM_001297647.1 | NP_001284576.1 | ||
| NM_001297648.1 | NP_001284577.1 | |||
| NM_006314.2 | NP_006305.2 | |||
| XM_011540484.2 | XP_011538786.1 | |||
| XM_011540485.2 | XP_011538787.1 | |||
| AB209335.1 | BAD92572.1 | |||
| AF100153.1 | AAC80558.1 | |||
| AK001900.1 | ||||
| AK222817.1 | BAD96537.1 | |||
| AK222904.1 | BAD96624.1 | |||
| AK292646.1 | ||||
| BC011604.2 | AAH11604.1 | |||
| BC012797.2 | AAH12797.1 | |||
| BT006900.1 | AAP35546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002191 | Chr.1:26149030 - 26219160 on Build GRCh38 | Gain |
|
| esv3577792 | Chr.1:26184821 - 26198215 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map